Learn More
Biomatik Corporation Mouse Edn1 (Endothelin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58082
Encompass_Preferred
Mouse Edn1 (Endothelin-1) ELISA Kit for detecting Edn1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80pg/ml. Sensitivity: 0.625pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22387. Target Synonyms: ET-1; Endothelin-1; EDN1; ET1; HDLCQ7; PPET1; Preproendothelin-1; endothelin 1; preproendothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5528