Learn More
Biomatik Corporation Mouse IL-16 (Interleukin 16) ELISA Kit
Supplier:  Biomatik Corporation EKF58085
Encompass_Preferred
Mouse IL-16 (Interleukin 16) ELISA Kit for detecting IL-16 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O54824. Target Synonyms: IL-16; Interleukin 16; IL16; LCF; NIL16; PRIL16; prIL-16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5530