Learn More
Biomatik Corporation Rat CREB (Cyclic AMP Response Element Binding Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58094
Encompass_Preferred
Rat CREB (Cyclic AMP Response Element Binding Protein) ELISA Kit for detecting CREB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15337. Target Synonyms: CREB; active transcription factor CREB; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB-1; cyclic AMP-responsive element-binding protein 1; MGC9284; transactivator protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5536