Learn More
Biomatik Corporation Rat Nkap (NF-kappa-B-activating protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58097
Encompass_Preferred
Rat Nkap (NF-kappa-B-activating protein) ELISA Kit for detecting Nkap in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q4V7C9. Target Synonyms: Nkap. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5538