Learn More
Biomatik Corporation Mouse Smad7 (Mothers against decapentaplegic homolog 7) ELISA Kit
Supplier:  Biomatik Corporation EKF58119
Encompass_Preferred
Mouse Smad7 (Mothers against decapentaplegic homolog 7) ELISA Kit for detecting Smad7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35253. Target Synonyms: Smad7; MADH7; Mothers Against Decapentaplegic Homolog 7; MADH8; CRCS3; hSMAD7; MAD homolog 7; MAD homolog 8; MAD; mothers against decapentaplegic homolog 7; Drosophila; Mothers against decapentaplegic homolog 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5557