Learn More
Biomatik Corporation Rat Gc (Glucagon) ELISA Kit
Supplier:  Biomatik Corporation EKF58121
Encompass_Preferred
Rat Gc (Glucagon) ELISA Kit for detecting Gc in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06883. Target Synonyms: GcG; Glucagon; GLP1; GLP2; GRPP; glicentin-related polypeptide; glucagon-like peptide 1; glucagon-like peptide 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5559