Learn More
Biomatik Corporation Mouse CaN (Calcineurin) ELISA Kit
Supplier:  Biomatik Corporation EKF58123
Encompass_Preferred
Mouse CaN (Calcineurin) ELISA Kit for detecting CaN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 19.531-1250pg/ml. Sensitivity: 11.719pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P63328. Target Synonyms: CaN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5561