Learn More
Biomatik Corporation Mouse Cd74 (HLA class II histocompatibility antigen gamma chain) ELISA Kit
Supplier:  Biomatik Corporation EKF58154
Encompass_Preferred
Mouse Cd74 (HLA class II histocompatibility antigen gamma chain) ELISA Kit for detecting Cd74 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04441. Target Synonyms: CD74; HLA class II histocompatibility antigen gamma chain; CLIP; DHLAG; HLADG; Ii; II; MHC HLA-DR gamma chain; p33; Ia-gamma; INVG34; CD74 antigen; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; Ia antigen-associated invariant chain; Ia-associated invariant chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5581