Learn More
Biomatik Corporation Rat TAT (Thrombin-Antithrombin Complex) ELISA Kit
Supplier:  Biomatik Corporation EKF58168
Encompass_Preferred
Rat TAT (Thrombin-Antithrombin Complex) ELISA Kit for detecting TAT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P18292; F7EY53. Target Synonyms: TAT; Thrombin; Antithrombin Complex; TATC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5592