Learn More
Biomatik Corporation Human CALR (Calreticulin) ELISA Kit
Supplier:  Biomatik Corporation EKF58187
Encompass_Preferred
Human CALR (Calreticulin) ELISA Kit for detecting CALR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27797. Target Synonyms: CALR; CRT; Calreticulin; grp60; HACBP; Endoplasmic reticulum resident protein60; ERp60; CRP55; CRTC; calregulin; CaBP3; calsequestrin-like protein; endoplasmic reticulum resident protein 60. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5604