Learn More
Biomatik Corporation Human LRP8 (Low-density lipoprotein receptor-related protein 8) ELISA Kit
Supplier:  Biomatik Corporation EKF58191
Encompass_Preferred
Human LRP8 (Low-density lipoprotein receptor-related protein 8) ELISA Kit for detecting LRP8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14114. Target Synonyms: LRP8; Low-density lipoprotein receptor-related protein 8; APOER2; LRP-8; Apolipoprotein E receptor 2; Lr8b; MCI1; Apolipoprotein E R2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5608