Learn More
Biomatik Corporation Rat PTX3 (Pentraxin 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58193
Encompass_Preferred
Rat PTX3 (Pentraxin 3) ELISA Kit for detecting PTX3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: D3ZT94. Target Synonyms: PTX3; TSG-14; alpha-induced protein 5; pentaxin-related gene; rapidly induced by IL-1 beta; tumor necrosis factor; Pentaxin-related protein PTX3; pentraxin 3; long; pentraxin-3; pentraxin-related gene; rapidly induced by IL-1 beta; pentraxin-related protein PTX3; TNF alpha-induced protein 5; TNFAIP5; TSG14; TSG-14pentaxin-related gene; rapidly induced by IL-1 beta; tumor necrosis factor; alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5610