Learn More
Biomatik Corporation Rat AMY2 (Amylase Alpha 2, Pancreatic) ELISA Kit
Supplier:  Biomatik Corporation EKF58200
Encompass_Preferred
Rat AMY2 (Amylase Alpha 2, Pancreatic) ELISA Kit for detecting AMY2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00689. Target Synonyms: Amy2; PA; AMY2; Amy2a; Pancreatic alpha-amylase; 1; 4-alpha-D-glucan glucanohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5614