Learn More
Biomatik Corporation Mouse GAL4 (Galectin 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58206
Encompass_Preferred
Mouse GAL4 (Galectin 4) ELISA Kit for detecting GAL4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8K419. Target Synonyms: Galectin-4; GAL4; Galectin 4; LGALS4; Antigen NY-CO-27; gal-4; Gal-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5618