Learn More
Biomatik Corporation Human RTN3 (Reticulon-3) ELISA Kit
Supplier:  Biomatik Corporation EKF58226
Encompass_Preferred
Human RTN3 (Reticulon-3) ELISA Kit for detecting RTN3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95197. Target Synonyms: ASYIP; HAP; NSP like protein 2; NSP like protein II; NSPL2; NSPLII; reticulon 3; RTN3; RTN3 A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5632