Learn More
Biomatik Corporation Human Foxj1 (Forkhead box protein J1) ELISA Kit
Supplier:  Biomatik Corporation EKF58235
Encompass_Preferred
Human Foxj1 (Forkhead box protein J1) ELISA Kit for detecting Foxj1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92949. Target Synonyms: Foxj1; FKHL13; HFH-4; HFH4; fork head homologue 4; forkhead transcription factor HFH-4; forkhead-like 13; forkhead-related protein FKHL13; hepatocyte nuclear factor 3 forkhead homolog 4; forkhead box J1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5638