Learn More
Biomatik Corporation Mouse GATA3 (GATA Binding Protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58243
Encompass_Preferred
Mouse GATA3 (GATA Binding Protein 3) ELISA Kit for detecting GATA3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23772. Target Synonyms: GATA binding protein 3; GATA-binding factor 3; GATA-binding protein 3; HDRMGC5199; MGC2346; MGC5445; trans-acting T-cell-specific transcription factor GATA-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5644