Learn More
Biomatik Corporation Human APOL1 (Apolipoprotein L1) ELISA Kit
Supplier:  Biomatik Corporation EKF58262
Encompass_Preferred
Human APOL1 (Apolipoprotein L1) ELISA Kit for detecting APOL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O14791. Target Synonyms: APOL1; Apolipoprotein L1; APOL; Apo-L; ApoL-I; Apolipoprotein L; Apolipoprotein L-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5660