Learn More
Biomatik Corporation Mouse Bcl2 (Apoptosis regulator Bcl-2) ELISA Kit
Supplier:  Biomatik Corporation EKF58270
Encompass_Preferred
Mouse Bcl2 (Apoptosis regulator Bcl-2) ELISA Kit for detecting Bcl2 in cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10417. Target Synonyms: Bcl2; Bcl-2; apoptosis regulator Bcl-2; B-cell CLL; lymphoma 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5665