Learn More
Biomatik Corporation Human WNT7B (Protein Wnt-7b) ELISA Kit
Supplier:  Biomatik Corporation EKF58280
Encompass_Preferred
Human WNT7B (Protein Wnt-7b) ELISA Kit for detecting WNT7B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P56706. Target Synonyms: WNT7B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5672