Learn More
Biomatik Corporation Human HLA-DQB2 (HLA class II histocompatibility antigen, DQ beta 2 chain) ELISA Kit
Supplier:  Biomatik Corporation EKF58288
Encompass_Preferred
Human HLA-DQB2 (HLA class II histocompatibility antigen, DQ beta 2 chain) ELISA Kit for detecting HLA-DQB2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05538. Target Synonyms: HLA DQB1; HLA DQB2; HLA DXB; HLA-DQB2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5676