Learn More
Biomatik Corporation Mouse GAL8 (Galectin 8) ELISA Kit
Supplier:  Biomatik Corporation EKF58306
Encompass_Preferred
Mouse GAL8 (Galectin 8) ELISA Kit for detecting GAL8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JL15. Target Synonyms: GAL8; LGALS8; PCTA1; Gal-8; galectin 8; galectin-8; galectin-8g; lectin; galactoside-binding; soluble; 8; PCTA1; PCTA-1Po66 carbohydRate-binding protein; Po66 carbohydRate binding protein; Po66-CBP; Prostate carcinoma tumor antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5686