Learn More
Biomatik Corporation Mouse Notch-1 (Neurogenic locus notch homolog protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58326
Encompass_Preferred
Mouse Notch-1 (Neurogenic locus notch homolog protein 1) ELISA Kit for detecting Notch-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q01705. Target Synonyms: Notch-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5702