Learn More
Biomatik Corporation Mouse FLT3 (Receptor-type tyrosine-protein kinase FLT3) ELISA Kit
Supplier:  Biomatik Corporation EKF58330
Encompass_Preferred
Mouse FLT3 (Receptor-type tyrosine-protein kinase FLT3) ELISA Kit for detecting FLT3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q00342. Target Synonyms: Flt3; FMS Like Tyrosine Kinase 3; CD135; Flk-2; STK-1; CD135; CD135 antigen; fetal liver kinase 2; FL cytokine receptor; FLK2; FLT-3; FLT3 receptor tyrosine kinase; Fms-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Stem cell tyrosine kinase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5705