Learn More
Biomatik Corporation Human IDH1 (IsocitRate dehydrogenase [NADP] cytoplasmic) ELISA Kit
Supplier:  Biomatik Corporation EKF58380
Encompass_Preferred
Human IDH1 (IsocitRate dehydrogenase [NADP] cytoplasmic) ELISA Kit for detecting IDH1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O75874. Target Synonyms: IDH1; IsocitRate dehydrogenase [NADP] cytoplasmic; Oxalosuccinate decarboxylase; NADP; +-specific ICDH; IDH; Cytosolic NADP-isocitRate dehydrogenase; IDP; PICD; IDCD; IDPC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5739