Learn More
Biomatik Corporation Human MBOAT4 (Ghrelin O-acyltransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF58385
Encompass_Preferred
Human MBOAT4 (Ghrelin O-acyltransferase) ELISA Kit for detecting MBOAT4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96T53. Target Synonyms: MBOAT4; OACT4; EC 2.3.1.-; ghrelin O-acyltransferase; membrane bound O-acyltransferase domain containing 4; Membrane-bound O-acyltransferase domain-containing protein 4; OACT4; O-acyltransferase; membrane bound domain containing 4; O-acyltransferase domain-containing protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5742