Learn More
Biomatik Corporation Human EGFR (Epidermal growth factor receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF58409
Encompass_Preferred
Human EGFR (Epidermal growth factor receptor) ELISA Kit for detecting EGFR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00533. Target Synonyms: EGFR; ErbB-1; ErbB1; ERBB; HER1; Mena. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5759