Learn More
Biomatik Corporation Rat FDP (Fibrinogen Degradation Product) ELISA Kit
Supplier:  Biomatik Corporation EKF58432
Encompass_Preferred
Rat FDP (Fibrinogen Degradation Product) ELISA Kit for detecting FDP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Target Synonyms: FDP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5772