Learn More
Biomatik Corporation Rat MT-1 (Metallothionein-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58471
Encompass_Preferred
Rat MT-1 (Metallothionein-1) ELISA Kit for detecting MT-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02803. Target Synonyms: MT-1; MT; MT1a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5800