Learn More
Biomatik Corporation Human PROC (Vitamin K-dependent protein C) ELISA Kit
Supplier:  Biomatik Corporation EKF58473
Encompass_Preferred
Human PROC (Vitamin K-dependent protein C) ELISA Kit for detecting PROC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04070. Target Synonyms: PROC; Protein C; Autoprothrombin IIA; Protein C; Anticoagulant protein C; APC; Blood coagulation factor XIV; PC; PROC1; protein C; inactivator of coagulation factors Va and VIIIa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5802