Learn More
Biomatik Corporation Mouse IL-12 (Interleukin 12) ELISA Kit
Supplier:  Biomatik Corporation EKF58493
Encompass_Preferred
Mouse IL-12 (Interleukin 12) ELISA Kit for detecting IL-12 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P43431; P43432. Target Synonyms: Interleukin 12 p70; CLMF p35; Cytotoxic lymphocyte matuRation factor 35 kDa subunit; IL-12 subunit p35; IL-12; subunit p35; interleukin 12; p35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5815