Learn More
Biomatik Corporation Human Leucine-rich repeat-containing protein 17 (LRRC17) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34406
Encompass_Preferred
The Human Leucine-rich repeat-containing protein 17 (LRRC17) ELISA Kit allows for quantitative determination of leucine rich repeat containing 17 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 23.44 pg/mL-1500 pg/mL. Sensitivity: 5.86 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8N6Y2. Restriction: For Research Use Only.
Catalog No. 50-149-8034