Learn More
Biomatik Corporation Mouse Hrg (Histidine-rich glycoprotein) ELISA Kit
Supplier:  Biomatik Corporation EKF58498
Encompass_Preferred
Mouse Hrg (Histidine-rich glycoprotein) ELISA Kit for detecting Hrg in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9ESB3. Target Synonyms: Hrg; HRG; HPRG; HRGP; THPH11; Histidine-proline-rich glycoprotein; HPRG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5818