Learn More
Biomatik Corporation Mouse TFF2 (Trefoil Factor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58501
Encompass_Preferred
Mouse TFF2 (Trefoil Factor 2) ELISA Kit for detecting TFF2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q03404. Target Synonyms: TFF2; SML1; SP; Spasmolysin; Spasmolytic Polypeptide; SML1trefoil factor 2; SML1; Human spasmolytic polypeptide; SP10spasmolytic protein 1; SP; spasmolysin; Spasmolysin; Spasmolytic polypeptide; trefoil factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5821