Learn More
Biomatik Corporation Human LILRB4 (Leukocyte immunoglobulin-like receptor subfamily B member 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58503
Encompass_Preferred
Human LILRB4 (Leukocyte immunoglobulin-like receptor subfamily B member 4) ELISA Kit for detecting LILRB4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NHJ6. Target Synonyms: LILRB4; CD85k; HM18; ILT3; LIR5; CD85 antigen-like family member K; CD85k; CD85k antigen; ILT-3; ILT3CD85K; leukocyte immunoglobulin-like receptor; subfamily B; with TM and ITIM domains; LIR5LILRB5; LIR-5subfamily B; member 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5823