Learn More
Biomatik Corporation Mouse PAI-2 (Plasminogen Activator Inhibitor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58516
Encompass_Preferred
Mouse PAI-2 (Plasminogen Activator Inhibitor 2) ELISA Kit for detecting PAI-2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12388. Target Synonyms: PAI-2; PLANH2; Urokinase Inhibitor; HsT1201; Monocyte Arg-serpin; PAI; PAI2Urokinase inhibitor; Placental plasminogen activator inhibitor; PLANH2PAI-2; plasminogen activator inhibitor 2; plasminogen activator inhibitor; type II; arginine-serpin; serine; or cysteine proteinase inhibitor; clade B; ovalbumin; member 2; Serpin B2; serpin peptidase inhibitor; clade B; ovalbumin; member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5832