Learn More
Biomatik Corporation Rat β-CTx (Beta Crosslaps) ELISA Kit
Supplier:  Biomatik Corporation EKF58531
Encompass_Preferred
Rat β-CTx (Beta Crosslaps) ELISA Kit for detecting β-CTx in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P02454; P02466. Target Synonyms: β-CTx. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5845