Learn More
Biomatik Corporation Human FCN3 (Ficolin-3) ELISA Kit
Supplier:  Biomatik Corporation EKF58536
Encompass_Preferred
Human FCN3 (Ficolin-3) ELISA Kit for detecting FCN3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O75636. Target Synonyms: Ficolin-3; FCN3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5850