Learn More
Biomatik Corporation Rat MIP-1β (Macrophage Inflammatory Protein 1 Beta) ELISA Kit
Supplier:  Biomatik Corporation EKF58542
Encompass_Preferred
Rat MIP-1β (Macrophage Inflammatory Protein 1 Beta) ELISA Kit for detecting MIP-1β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100pg/ml. Sensitivity: 0.938pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50230. Target Synonyms: CCL4; MIP-1β; ACT2; AT744.1; G-26; HC21; LAG-1; LAG1; MIP-1-beta; MIP1B1; SCYA2; SCYA4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5855