Learn More
Biomatik Corporation Mouse IL-33 (Interleukin 33) ELISA Kit
Supplier:  Biomatik Corporation EKF58546
Encompass_Preferred
Mouse IL-33 (Interleukin 33) ELISA Kit for detecting IL-33 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8BVZ5. Target Synonyms: IL-33; C9orf26; DVS27; NF-HEV; chromosome 9 open reading frame 26; NF-HEV; DKFZp586H0523; DVS27-related protein; IL1F11; IL-1F11; IL-33; interleukin 33; Interleukin-1 family member 11; interleukin-33; NFHEV; NF-HEVNFEHEV; Nuclear factor from high endothelial venules; RP11-575C20.2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5857