Learn More
Biomatik Corporation Human NFATC1 (Nuclear factor of activated T-cells, cytoplasmic 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58553
Encompass_Preferred
Human NFATC1 (Nuclear factor of activated T-cells, cytoplasmic 1) ELISA Kit for detecting NFATC1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95644. Target Synonyms: NFATC1; NFAT2; NFAT transcription complex cytosolic component; NFATC; NF-ATC; NF-ATc1; nuclear factor of activated T-cells; cytoplasmic; calcineurin-dependent 1; Nuclear Factor of Activated T Cells C1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5864