Learn More
Biomatik Corporation Human PLAC8 (Placenta-specific gene 8 protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58564
Encompass_Preferred
Human PLAC8 (Placenta-specific gene 8 protein) ELISA Kit for detecting PLAC8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NZF1. Target Synonyms: C15; onzin; PLAC8; placenta 8; Protein C15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5871