Learn More
Biomatik Corporation Rat Pnlip (Pancreatic triacylglycerol lipase) ELISA Kit
Supplier:  Biomatik Corporation EKF58574
Encompass_Preferred
Rat Pnlip (Pancreatic triacylglycerol lipase) ELISA Kit for detecting Pnlip in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27657. Target Synonyms: Pnlip; Pancreatic triacylglycerol lipase; PL; PTL; Pancreatic lipase; PNLIPD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5879