Learn More
Biomatik Corporation Rat Reg3b (RegeneRating islet-derived protein 3-beta) ELISA Kit
Supplier:  Biomatik Corporation EKF58623
Encompass_Preferred
Rat Reg3b (RegeneRating islet-derived protein 3-beta) ELISA Kit for detecting Reg3b in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P25031. Target Synonyms: Reg3b; Reg3β; HIP; PAP1; Pap; REG-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5916