Learn More
Biomatik Corporation Human SERPINB3 (Serpin B3) ELISA Kit
Supplier:  Biomatik Corporation EKF58644
Encompass_Preferred
Human SERPINB3 (Serpin B3) ELISA Kit for detecting SERPINB3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P29508. Target Synonyms: SCCA1; Squamous Cell Carcinoma Antigen 1SERPINB3; T4-A; HsT1196; Protein T4-A; SCC; SCCA; SCCA-1; SCCA-PD; serpin B3; serpin peptidase inhibitor; clade B; ovalbumin; member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5933