Learn More
Biomatik Corporation Mouse Tshβ (Thyrotropin subunit beta) ELISA Kit
Supplier:  Biomatik Corporation EKF58649
Encompass_Preferred
Mouse Tshβ (Thyrotropin subunit beta) ELISA Kit for detecting Tshβ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10uiu/ml. Sensitivity: 0.094uiu/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P12656. Target Synonyms: Tshβ; TSHB; Thyrotropin subunit beta; Thyrotropin beta; thyroid stimulating hormone; beta; thyroid-stimulating hormone subunit beta; thyrotropin beta chain; thyrotropin beta subunit; TSH-B; TSH-BETA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5936