Learn More
Biomatik Corporation Human TFPI (Tissue factor pathway inhibitor) ELISA Kit
Supplier:  Biomatik Corporation EKF58658
Encompass_Preferred
Human TFPI (Tissue factor pathway inhibitor) ELISA Kit for detecting TFPI in plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10646. Target Synonyms: TFPI; Tissue Factor Pathway Inhibitor; EPI; LACI; TFI; TFPI1; lipoprotein-associated coagulation inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5944