Learn More
Biomatik Corporation Human GROγ (Growth Regulated Oncogene Gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF58672
Encompass_Preferred
Human GROγ (Growth Regulated Oncogene Gamma) ELISA Kit for detecting GROγ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19876. Target Synonyms: GROγ; CXCL3; CINC-2; DCIP-1; GRO gamma; Dcip1; CINC-2b; GRO3; GRO3 oncogene; GROG; GRO-gamma; MIP2B; MIP-2b; MIP2-beta; C-X-C motif chemokine 3; Growth-regulated protein gamma; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5954