Learn More
Biomatik Corporation Mouse CCL24 (C-C motif chemokine 24) ELISA Kit
Supplier:  Biomatik Corporation EKF58695
Encompass_Preferred
Mouse CCL24 (C-C motif chemokine 24) ELISA Kit for detecting CCL24 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JKC0. Target Synonyms: MPIF2; MPIF-2; CCL24; Eotaxin-2; chemokine; C-C motif ligand 24; Ckb-6; Eosinophil chemotactic protein 2; Eotaxin-2; member 24; MPIF2Myeloid progenitor inhibitory factor 2; MPIF-2Small-inducible cytokine A24; SCYA24CK-beta-6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5971