Learn More
Biomatik Corporation Human PFN1 (Profilin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58697
Encompass_Preferred
Human PFN1 (Profilin-1) ELISA Kit for detecting PFN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07737. Target Synonyms: PFN1; Profilin-1; Profilin I; PROF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5973